Learn More
Qc Supply LLC Virkon S Dilution Test Strips, 60/pk
SDP
Catalog No. NC1807436
Encompass
Accurately validate you have made the correct concentration of diluted Virkon S solution with Virkon S Dilution Test Strips. This technical tool will help validate the pre-measured and diluted solution is at the appropriate concentration to provide the efficacy needed to combat the full list of pathogens Virkon S protects against.
Catalog No. NC1807436 Supplier Qc Supply LLC Supplier No. 340476
Accurately validate you have made the correct concentration of diluted Virkon S solution with Virkon S Dilution Test Strips. This technical tool will help validate the pre-measured and diluted solution is at the appropriate concentration to provide the efficacy needed to combat the full list of pathogens Virkon S protects against.