Learn More
International Enviroguard™ ViroGuard™ 2404 Disposable Coveralls with Hood and Boot
Catalog No. 17100881 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-100-881 X-Large
17-100-882 Medium
17-100-879 Large
17-100-877 2X-Large
17-100-880 3X-Large
17-100-883 4X-Large
17-100-878 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-100-881 Supplier International Enviroguard™ Supplier No. 2404XL
Add to Cart
Edge
Add to Cart
ViroGuard™ Disposable Coveralls are designed and tested to protect against blood, blood-borne pathogens, bodily fluids, and many other liquid hazards. These coveralls boast elastic wrists and back, and a front zipper with storm flap.
  • Tear- and abrasion-resistant fabric protects against scrapes and scratches in the toughest of work environments
  • Fabric helps prevent heat stress with enhanced moisture vapor transmission, so you stay cool and comfortable on the job
  • With an attached hood and boots, you're protected from head-to-toe
  • Blue color makes stains and splashes less conspicuous
  • Possible applications include emergency response, category 3 black water, tank cleaning, crime scene remediation, and animal research
  • Fabric passes ASTM F1670 and F1671 tests for protection against blood, bodily fluids and blood borne pathogens
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure area
  • Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric
  • Elastic back stretches with user for more comfort and better tear resistance
Specifications
Product Type Protective Coveralls
Size X-Large
Gender Unisex
Material Microporous Laminate
Color Blue
Disposable Single-use
Waist Style Loose
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Class 4
Resistance Bloodborne Pathogens, Liquid, Chemicals, Tears, Abrasion