Learn More
ATCC Virome -; Nucleic Acid Mix
Catalog No. 502385067
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises an even mixture of nucleic acids prepared from fully sequenced, characterized, and authenticated viral strains selected on the basis of genomic size, DNA/RNA genome, envelope/non-envelope, and other special features. Specification Range: 2x104 genome copies/μL ± 1 log* *Nucleic acid concentrations indicate ATCC manufacturing specifications and are provided as a reference only
Catalog No. 50-238-5067 Supplier ATCC Supplier No. MSA1008
Add to Cart
Add to Cart
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises an even mixture of nucleic acids prepared from fully sequenced, characterized, and authenticated viral strains selected on the basis of genomic size, DNA/RNA genome, envelope/non-envelope, and other special features. Specification Range: 2x104 genome copies/μL ± 1 log* *Nucleic acid concentrations indicate ATCC manufacturing specifications and are provided as a reference only