Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions Virtu-Clean 70% Isopropyl Alcohol

Catalog No. 17377753
Change view
Click to view available options
Scent:
Unscented
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scent
17-377-753 Unscented
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-377-753 Supplier High-Tech Conversions Supplier No. HBG703016797
Add to Cart
Add to Cart

Virtu-Clean™ Sterile Alcohol is 70% isopropyl alcohol / 30% water for injection cleaner for critical environments. This IPA is filtered at 0.1 microns which is less than the 0.22 industry standard for added cleanliness.

Virtu-Clean™ Sterile Alcohol is a 70% Isopropyl Alcohol / 30% Water for Injection cleaner for critical environments. This IPA is filtered at 0.1 microns which is less than the 0.22 industry standard for added cleanliness.

  • Validated sterile
  • Available in 16 oz. trigger spray bottle
  • Removes many surface contaminants
  • Used for wiping down gloved hands, vials, equipment, supplies, counter tops, floors, walls and more
  • Double bagged
  • ISO class 5
  • USP <797> and USP <800> compliant

Specifications

Product Type Sterile IPA
Color-coded No
Form Liquid Spray
pH 6.0 to 8.0
Scent Unscented
Sterility Sterile
Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
Filtered Filtered
Flammable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.