Learn More
MSC VIRTUA AP SAFETY GLASSES
Catalog No. 5000767293
Encompass
These economy-rated safety glasses combine style, fit, lightweight comfort, and affordability. They feature integrated side shields for additional protection and a wraparound design that offers extended eye protection and unobstructed viewing. The contoured temples are designed for a comfortable and snug fit. The polycarbonate lenses absorb 99.9% of UVA and UVB rays.
  • Integrated side shields provide additional protection
  • Wraparound design provides extended eye protection and unobstructed viewing
  • Contoured temples fit comfortably and snugly
  • Polycarbonate lenses absorb 99.9% UVA and UVB
  • Anti-fog and scratch-resistant coatings improve visibility
  • Meets ANSI Z87.1-2015 standards
Catalog No. 50-007-67293 Supplier MSC Supplier No. 64556988
These economy-rated safety glasses combine style, fit, lightweight comfort, and affordability. They feature integrated side shields for additional protection and a wraparound design that offers extended eye protection and unobstructed viewing. The contoured temples are designed for a comfortable and snug fit. The polycarbonate lenses absorb 99.9% of UVA and UVB rays.
  • Integrated side shields provide additional protection
  • Wraparound design provides extended eye protection and unobstructed viewing
  • Contoured temples fit comfortably and snugly
  • Polycarbonate lenses absorb 99.9% UVA and UVB
  • Anti-fog and scratch-resistant coatings improve visibility
  • Meets ANSI Z87.1-2015 standards