Learn More
MSC VIRTUA AP SAFETY GLASSES
Catalog No. 5000767055
Encompass
Economy-rated safety glasses that combine style, fit, lightweight comfort, and affordability. They feature integrated side-shields for additional protection and a wraparound design that offers extended eye protection and unobstructed viewing. The contoured temples are designed for a comfortable and snug fit. The polycarbonate lenses absorb 99.9% of UVA and UVB rays.
  • Integrated side-shields provide additional protection
  • Wraparound design provides extended eye protection and unobstructed viewing
  • Contoured temples fit comfortably and snugly
  • Polycarbonate lenses absorb 99.9% UVA and UVB
  • Scratch-resistant lens coating
  • Frameless design with clear frame and temples
  • Meets ANSI Z87.1-2015 standards
  • Universal size
Catalog No. 50-007-67055 Supplier MSC Supplier No. 64556905
Economy-rated safety glasses that combine style, fit, lightweight comfort, and affordability. They feature integrated side-shields for additional protection and a wraparound design that offers extended eye protection and unobstructed viewing. The contoured temples are designed for a comfortable and snug fit. The polycarbonate lenses absorb 99.9% of UVA and UVB rays.
  • Integrated side-shields provide additional protection
  • Wraparound design provides extended eye protection and unobstructed viewing
  • Contoured temples fit comfortably and snugly
  • Polycarbonate lenses absorb 99.9% UVA and UVB
  • Scratch-resistant lens coating
  • Frameless design with clear frame and temples
  • Meets ANSI Z87.1-2015 standards
  • Universal size