Learn More
MSC VIRTUA AP SAFETY GLASSES
Catalog No. 5000792943
Encompass
These safety glasses offer a blend of style, comfort, and affordability, featuring a lightweight, wrap-around design for extended eye protection and an unobstructed field of vision. Equipped with polycarbonate lenses, they provide integrated side shields and absorb 99.9% of UVA and UVB radiation.
  • Optical Class 1 for prolonged comfortable wear
  • Scratch-resistant lens coating
  • Integrated side shields provide additional protection
  • Wrap-around design provides excellent coverage
  • Polycarbonate lenses absorb 99.9% UVA and UVB
  • Light blue lens minimizes glare, eye stress, and fatigue
  • Lightweight (26g) for maximum comfort
  • Meets ANSI Z87.1-2015 standards
Catalog No. 50-007-92943 Supplier MSC Supplier No. 64556947
These safety glasses offer a blend of style, comfort, and affordability, featuring a lightweight, wrap-around design for extended eye protection and an unobstructed field of vision. Equipped with polycarbonate lenses, they provide integrated side shields and absorb 99.9% of UVA and UVB radiation.
  • Optical Class 1 for prolonged comfortable wear
  • Scratch-resistant lens coating
  • Integrated side shields provide additional protection
  • Wrap-around design provides excellent coverage
  • Polycarbonate lenses absorb 99.9% UVA and UVB
  • Light blue lens minimizes glare, eye stress, and fatigue
  • Lightweight (26g) for maximum comfort
  • Meets ANSI Z87.1-2015 standards