Learn More
MSC VIRTUA SAFETY GLASSES
Catalog No. 5000685838
Encompass
These safety glasses provide essential eye protection with clear, frameless polycarbonate lenses treated for anti-fog and scratch resistance. Featuring a wrap-around design and fixed temples, they offer universal fit and UV protection, complying with ANSI Z87.1-2015 standards.
  • Clear, frameless polycarbonate lenses
  • Anti-fog coating for enhanced visibility
  • Scratch-resistant lenses for durability
  • Wrap-around design with fixed temples
  • Universal size for broad applicability
  • UV protection provided
  • Meets ANSI Z87.1-2015 safety standards
Catalog No. 50-006-85838 Supplier MSC Supplier No. 62946165
These safety glasses provide essential eye protection with clear, frameless polycarbonate lenses treated for anti-fog and scratch resistance. Featuring a wrap-around design and fixed temples, they offer universal fit and UV protection, complying with ANSI Z87.1-2015 standards.
  • Clear, frameless polycarbonate lenses
  • Anti-fog coating for enhanced visibility
  • Scratch-resistant lenses for durability
  • Wrap-around design with fixed temples
  • Universal size for broad applicability
  • UV protection provided
  • Meets ANSI Z87.1-2015 safety standards