Learn More
Med Vet International Miltex Vantage Virtus Splinter Forceps, 6in.
Catalog No. 502831051
Encompass_Preferred
The Miltex V96-336 Virtus Splinter Forceps are made of mid-grade stainless steel and have a length of 6in. They have a non-locking spring handle with a pointed serrated tip.
Catalog No. 50-283-1051 Supplier Med Vet International Supplier No. MILV96336
The Miltex V96-336 Virtus Splinter Forceps are made of mid-grade stainless steel and have a length of 6in. They have a non-locking spring handle with a pointed serrated tip.