Learn More
MSC Rochester Midland Corporation Sanitizer, Concentrated Liquid, Quaternary Ammonium, Food Contact, No Rinse
Catalog No. 502839966
Encompass
F-29 is a concentrated, cost-effective liquid sanitizer formulated with a blend of four quaternary ammonium chlorides. It is highly effective in controlling microorganisms on hard food contact surfaces. This sanitizer is hard water tolerant, non-corrosive on metal and plastic surfaces, and heat stable, making it suitable for various food processing plants including potato processors, bakeries, beverage, dairy, food, meat, and poultry.
  • Cost effective.
  • No potable water rinse required for pre-cleaned surfaces when used at specified dilutions.
  • Hard water tolerant up to 500 ppm hardness.
  • Non-corrosive on metal and plastic surfaces.
  • Effective against common foodborne pathogens.
  • Provides a residual bacteriostatic film on food contact surfaces.
  • Certified Kosher and phosphate free.
Catalog No. 50-283-9966 Supplier MSC Supplier No. 90358482
F-29 is a concentrated, cost-effective liquid sanitizer formulated with a blend of four quaternary ammonium chlorides. It is highly effective in controlling microorganisms on hard food contact surfaces. This sanitizer is hard water tolerant, non-corrosive on metal and plastic surfaces, and heat stable, making it suitable for various food processing plants including potato processors, bakeries, beverage, dairy, food, meat, and poultry.
  • Cost effective.
  • No potable water rinse required for pre-cleaned surfaces when used at specified dilutions.
  • Hard water tolerant up to 500 ppm hardness.
  • Non-corrosive on metal and plastic surfaces.
  • Effective against common foodborne pathogens.
  • Provides a residual bacteriostatic film on food contact surfaces.
  • Certified Kosher and phosphate free.