Learn More
Cayman Chemical Virus-URP6 (Bio-Rad) Multiplexed RT-qPCR Assay, 1 ea
SDP
Catalog No. 502945787
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X RT-qPCR enzyme mastermix Organism Type: Virus (-)ssRNA, Virus (+)ssRNA, Multiplexed Reaction Panels: COVID, Viral-URP Colors: Cys5 (Influenza B), FAM (Influenza A), HEX (RPP30), TEX615 (SARS-CoV2), TAMRA (Passive Reference) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-5787 Supplier Cayman Chemical Supplier No. 406991 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X RT-qPCR enzyme mastermix Organism Type: Virus (-)ssRNA, Virus (+)ssRNA, Multiplexed Reaction Panels: COVID, Viral-URP Colors: Cys5 (Influenza B), FAM (Influenza A), HEX (RPP30), TEX615 (SARS-CoV2), TAMRA (Passive Reference) Instruments: Bio-Rad CFX96 or CFX384