Learn More
Cayman Chemical VIrus-URP6MIC MultIplexed 1ea
SDP
Catalog No. 502945786
Encompass_Preferred
Oliogonucleotide recipes are optimized for BIS MIC. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X RT-qPCR enzyme mastermix Organism Type: Virus (-)ssRNA, Virus (+)ssRNA, Multiplexed Reaction Panels: COVID, Viral-URP Colors: Cys5 (Influenza B), FAM (Influenza A), HEX (RPP30), TexasRED615 (SARS-CoV2) Instruments: BMS MIC
Catalog No. 50-294-5786 Supplier Cayman Chemical Supplier No. 407011 EA
Add to Cart
Add to Cart
Oliogonucleotide recipes are optimized for BIS MIC. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X RT-qPCR enzyme mastermix Organism Type: Virus (-)ssRNA, Virus (+)ssRNA, Multiplexed Reaction Panels: COVID, Viral-URP Colors: Cys5 (Influenza B), FAM (Influenza A), HEX (RPP30), TexasRED615 (SARS-CoV2) Instruments: BMS MIC