Learn More
Cayman Chemical VIrus-URP6QuntStudIo Mult 1ea
SDP
Catalog No. 502945788
Encompass_Preferred
Oliogonucleotide recipes are optimized for QuantStudio 5, 6, 7, 12K. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X RT-qPCR enzyme mastermix Organism Type: Virus (-)ssRNA, Virus (+)ssRNA, Multiplexed Reaction Panels: COVID, Viral-URP Colors: Cys5 (Influenza B), FAM (Influenza A), HEX (RPP30), TEX615 (SARS-CoV2) Instruments: QuantStudio 5,6,7,12K
Catalog No. 50-294-5788 Supplier Cayman Chemical Supplier No. 407001 EA
Oliogonucleotide recipes are optimized for QuantStudio 5, 6, 7, 12K. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X RT-qPCR enzyme mastermix Organism Type: Virus (-)ssRNA, Virus (+)ssRNA, Multiplexed Reaction Panels: COVID, Viral-URP Colors: Cys5 (Influenza B), FAM (Influenza A), HEX (RPP30), TEX615 (SARS-CoV2) Instruments: QuantStudio 5,6,7,12K