Learn More
AdipoGen Virustomycin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20157M001
Encompass_Preferred
Chemical. CAS: 84777-85-5. Formula: C48H71NO14. MW: 886.1. Isolated from Streptomyces sp. AM-2604. Antibiotic. Anti-viral compound. Antifungal (weak). Anti-protozoal (Trypanosoma, Trichomonas).
Catalog No. 50-205-3542