Learn More
Azzota Corp Economic Digital Vis Spectrophotometer, Wavelength 325-1000Nm, 4Nm Bandwidth, 128 X 64 Lcd. Auto Wl Setting. Usb Connection. Can Display 15 Standard Curves & 50 Groups Of Data.
Catalog No. 507532958
Encompass_Preferred
Economic Digital VIS Spectrophotometer, Wavelength 325-1000nm, 4nm Bandwidth, 128 x 64 LCD. Auto WL Setting. USB connection. Can display 15 standard curves & 50 groups of data.
Catalog No. 50-753-2958 Supplier Azzota Corp Supplier No. SV1200
Economic Digital VIS Spectrophotometer, Wavelength 325-1000nm, 4nm Bandwidth, 128 x 64 LCD. Auto WL Setting. USB connection. Can display 15 standard curves & 50 groups of data.