Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves

Catalog No. 19048787 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-787 Large
19-048-786 Medium
19-048-785 Small
19-048-784 X-Small
19-048-788 X-Large
19-048-789 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-048-787 Supplier Wells Lamont™ Supplier No. Y9294L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves are constructed with a unique blend of high performance fibers and stainless steel to provide comfort and cut resistance. Polyurethane coating on the palm enhances grip capabilities when handling small parts.

  • Constructed from a blend of high performance fibers and stainless steel
  • 13-gauge shell provides ANSI Level A4 cut protection
  • Polyurethane coating on the palm enhances grip and tactility
  • Hi-vis coloring increases visibility during low light applications
  • Open, uncoated back circulates air to keep hands cool and comfortable

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance ASTM F1790-97, ASTM F2992-15, ASTM D3889-10
Coating Material Polyurethane
Color Hi-vis Orange, Black
Cut Resistance Rating ANSI A4
Hand Specific
Material HPPE, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4543
For Use With (Application) Assembly, construction, metal fabrication
Glove Cuff Style Knit
Model Y9294L
Series FlexTech™ Vis-Tech Y9294
Yarn Gauge 13
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.