Learn More
Aviva Systems Biology Corp VISA Antibody - N-terminal region (ARP49558_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49558_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VISA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VISA. Alternate names: CARDIF, DKFZp666M015, FLJ27482, FLJ41962, IPS-1, KIAA1271, MAVS, IPS1, VISAApplicationIHC, WB. Species reactivity: Cow, Human, Mouse
Catalog No. 50-014-60113