Learn More
Cliawaived Inc Sexual Health PCR Tests - Chlamydia/Gonorrhea/Trichomoniasis (Box of 20)
SDP
Catalog No. NC2578532
Encompass
The first true PCR point-of-care device for STI that’s all-in-one and requires no separate instrument or reader. Results available during the patient’s visit help to eliminate the guesswork of empirical medicine and patient callbacks associated with lab send outs. With the confidence of a PCR result, clinicians can prescribe the correct treatment and engage in a clear, guiding discussion with the patient. Note: Collection kits must be purchased seperately.Our sexual health test targets three pathogens that (Chlamydia/Gonorrhea/Trichomoniasis ), if left untreated, can cause permanent damage to a woman’s reproductive system, including infertility, or potentially causing fatal ectopic pregnancy (pregnancy that occurs outside the womb). Fast PCR can help with early detection of these serious infections.
Catalog No. NC2578532 Supplier Cliawaived Inc Supplier No. VSBPS00372
The first true PCR point-of-care device for STI that’s all-in-one and requires no separate instrument or reader. Results available during the patient’s visit help to eliminate the guesswork of empirical medicine and patient callbacks associated with lab send outs. With the confidence of a PCR result, clinicians can prescribe the correct treatment and engage in a clear, guiding discussion with the patient. Note: Collection kits must be purchased seperately.Our sexual health test targets three pathogens that (Chlamydia/Gonorrhea/Trichomoniasis ), if left untreated, can cause permanent damage to a woman’s reproductive system, including infertility, or potentially causing fatal ectopic pregnancy (pregnancy that occurs outside the womb). Fast PCR can help with early detection of these serious infections.