Learn More
Brookfield Engineer Labs General Purpose Oil, B600, Nominal Viscosity cP 600
Catalog No. NC0988613
Encompass
Accuracy: +/-1% of viscosity valueAppropriate for use at shear rates greater than 500 sec-1Recommended for use with cone/plate Viscometers at viscosities above 5,000 cPBrookfield oil viscosity standards are hydrocarbon based, either mineral oil or polybutenes Note: Other oil fluids are available - call for detailsBrookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify calibration of your instrument. All fluids are supplied in 1/2 liter (1 pint) containers complete with a certificate of calibration. CAP Oil Fluids are supplied in 150 mL (4 oz) containers
Catalog No. NC0988613 Supplier Brookfield Engineer Labs Supplier No. B600
Accuracy: +/-1% of viscosity valueAppropriate for use at shear rates greater than 500 sec-1Recommended for use with cone/plate Viscometers at viscosities above 5,000 cPBrookfield oil viscosity standards are hydrocarbon based, either mineral oil or polybutenes Note: Other oil fluids are available - call for detailsBrookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify calibration of your instrument. All fluids are supplied in 1/2 liter (1 pint) containers complete with a certificate of calibration. CAP Oil Fluids are supplied in 150 mL (4 oz) containers