Learn More
BURKLE INC ViscoDispo SteriPlast, HDPE sterile, 39.37 in. (1000 mm), 6.76 oz. (200 ml)
Catalog No. NC2187727
Encompass_Preferred
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100,000 mPas. The cleanroom-manufactured, individually packaged, gamma-ray sterilized design ensures safe and contamination-free sampling. The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.
This item is not returnable. View return policy
Catalog No. NC2187727 Supplier BURKLE INC Supplier No. 53933331
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100,000 mPas. The cleanroom-manufactured, individually packaged, gamma-ray sterilized design ensures safe and contamination-free sampling. The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.