Learn More
PCE AMERICAS INC Viscometer with a resolution 0.01 cP and repeatability ±0.5 %
SDP
Supplier:  PCE AMERICAS INC PCERVI 2
Encompass_Preferred
"Viscometer with LC display / Mains operation / Sample quantity 250 ... 400 ml /Measuring range 15 ... 100,000 cP / Adjustable speed of rotation /Simple operation / Spindle set L1 ... L4 included in delivery.The viscometer has a measuring range of 15 ... 100,000 cP. With an accuracy of <±2% of the range, the viscometer is a precise measuring device to determine the viscosity of a sample liquid. The viscosity is determined by the viscometer on the basis of an applied mechanical resistance that acts in the opposite direction to the rotational movement of a spindle. The viscometer can be operated with an adjustable speed in the range between 6… 60 RPM."
Catalog No. NC3654905