Learn More
PCE AMERICAS INC Viscometer with a repeatability of ±0.2% of the set measuring range
SDP
Catalog No. NC3654906
Encompass_Preferred
"The rotary viscometer uses the mechanical resistance of any liquid to measure viscosity using rotational movement of a spindle. This results in a torque that is recognized by the rotary viscometer and converted into a viscosity value. With an accuracy of ± 1.0%, the rotary viscometer is a very precise measuring device despite the high measuring range of 10 … 2,000,000 cP (mPa*s).Viscosity measuring range: 10 … 2,000,000 cPAccuracy: ± 1% of full scale (with Newtonian liquid)Rotation speeds: 0.3, 0.6, 1.5, 3, 6, 12, 30, 60 rpmVarious viscosity units can be displayed (mPa*s, Pa*s, dPa*s, cP, P, cPs)"
Catalog No. NC3654906 Supplier PCE AMERICAS INC Supplier No. PCERVI 10
"The rotary viscometer uses the mechanical resistance of any liquid to measure viscosity using rotational movement of a spindle. This results in a torque that is recognized by the rotary viscometer and converted into a viscosity value. With an accuracy of ± 1.0%, the rotary viscometer is a very precise measuring device despite the high measuring range of 10 … 2,000,000 cP (mPa*s).Viscosity measuring range: 10 … 2,000,000 cPAccuracy: ± 1% of full scale (with Newtonian liquid)Rotation speeds: 0.3, 0.6, 1.5, 3, 6, 12, 30, 60 rpmVarious viscosity units can be displayed (mPa*s, Pa*s, dPa*s, cP, P, cPs)"