Learn More
PCE AMERICAS INC Viscometer with a 5" LCD touchscreen that features resolution of 1 cp and a accuracy of ±1 % of full scale.
SDP
Catalog No. NC3654907
Encompass_Preferred
"The viscometer/viscosity meter has a measuring range of 27…. 5250 cp and an accuracy of +1% of the measuring range. The viscometer is used, for example, in quality control of production processes. In addition, the viscometer is particularly used in laboratories of all kinds.Measuring range: 27 … 5250 cpRotation speed: 200 RPMVolume measuring cup: 600 mlLarge touch screen for easy operationTemperature measuring range: -20 … 250 °C / -4 … 482 °F"
Catalog No. NC3654907 Supplier PCE AMERICAS INC Supplier No. PCERVI 8
"The viscometer/viscosity meter has a measuring range of 27…. 5250 cp and an accuracy of +1% of the measuring range. The viscometer is used, for example, in quality control of production processes. In addition, the viscometer is particularly used in laboratories of all kinds.Measuring range: 27 … 5250 cpRotation speed: 200 RPMVolume measuring cup: 600 mlLarge touch screen for easy operationTemperature measuring range: -20 … 250 °C / -4 … 482 °F"