Learn More
BURKLE INC ViscoSampler, PP, volume 5.41 oz. (160 ml), 23.62 in. (60 cm), Ø 1.26 in. (32 mm)
Catalog No. 5000026040
Encompass_Preferred
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Made of ultra-pure, chemically inert PTFE/FEP for contamination-free sampling.
Easy cleaning - all surfaces are poreless and without crevices. Dirt accumulation is prevented. Only round screw threads tried and tested in the field of food hygiene are used. Disassembly and cleaning are extremely simple.
Catalog No. 50-000-26040 Supplier BURKLE INC Supplier No. 53311060
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Made of ultra-pure, chemically inert PTFE/FEP for contamination-free sampling.
Easy cleaning - all surfaces are poreless and without crevices. Dirt accumulation is prevented. Only round screw threads tried and tested in the field of food hygiene are used. Disassembly and cleaning are extremely simple.