Learn More
BURKLE INC ViscoSampler, PTFE/FEP, volume 22.01 oz. (650 ml), 78.74 in. (200 cm), Ø 1.26 in. (32 mm)
Catalog No. 5000025876
Encompass_Preferred
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Made of ultra-pure, chemically inert PTFE/FEP for contamination-free sampling.
Easy cleaning - all surfaces are poreless and without crevices. Dirt accumulation is prevented. Only round screw threads tried and tested in the field of food hygiene are used. Disassembly and cleaning are extremely simple.
Catalog No. 50-000-25876 Supplier BURKLE INC Supplier No. 53312200
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Made of ultra-pure, chemically inert PTFE/FEP for contamination-free sampling.
Easy cleaning - all surfaces are poreless and without crevices. Dirt accumulation is prevented. Only round screw threads tried and tested in the field of food hygiene are used. Disassembly and cleaning are extremely simple.