Learn More
Canon Instrument Co * HAZARD * N1.0 Viscosity Standard 0.45 L
SDP
Catalog No. NC0187064
Encompass_Preferred
CANNON general purpose standards meet the precision specifications of ASTM D445/D446 and ISO 3104/3105. Lot specific data is provided with each standard.
Catalog No. NC0187064 Supplier Canon Instrument Co Supplier No. 9727C20.016
CANNON general purpose standards meet the precision specifications of ASTM D445/D446 and ISO 3104/3105. Lot specific data is provided with each standard.