Learn More
Canon Instrument Co N14 Viscosity Standard 0.5 L
SDP
Catalog No. NC0471695
Encompass_Preferred
CANNON general purpose standards meet the precision specifications of ASTM D445/D446 and ISO 3104/3105. Lot specific data is provided with each standard.
Catalog No. NC0471695 Supplier Canon Instrument Co Supplier No. 9727C34.016
CANNON general purpose standards meet the precision specifications of ASTM D445/D446 and ISO 3104/3105. Lot specific data is provided with each standard.