Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viscosity standard, Specpure™, nominally 500cSt @ 40 and 52cSt @ 100

Catalog No. aa47155ap
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47155AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA47155AP Supplier Thermo Scientific Chemicals Supplier No. 047155AP
Only null left
Add to Cart

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00084411
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Liquid
Quantity 500 mL
Grade Specpure
Chemical Name or Material Viscosity standard, Nominally 500cSt at 40 and 52cSt at 100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.