Learn More
Canon Instrument Co RT10 Silicone Oil Std 25C 0.5 L
SDP
Catalog No. NC1596243
Encompass_Preferred
CANNON silicone viscosity standards are specifically formulated for use with rotational viscometers. Lot specific viscosity data in mPa-s (cP) at 20 °C, 23 °C, 24 °C, 25 °C, 26 °C, 27 °C and 40 °C is provided with each standard.
Catalog No. NC1596243 Supplier Canon Instrument Co Supplier No. 9727Z14.016
CANNON silicone viscosity standards are specifically formulated for use with rotational viscometers. Lot specific viscosity data in mPa-s (cP) at 20 °C, 23 °C, 24 °C, 25 °C, 26 °C, 27 °C and 40 °C is provided with each standard.