Learn More
Brookfield Engineer Labs Brookfield 100 cP Viscosity Standard 25C 1000ml PB 100 CPS PB
Supplier:  Brookfield Engineer Labs 100 CPS/DUP
Encompass
General purpose silicone fluids are the most commonly used standard for verifying calibration of your Brookfield Ametek Viscometer/Rheometer.Silicone Standards are Newtonian fluids which are accurate to ±1% of viscosity value. Silicone fluids provide excellent temperature stability and are less temperature sensitive than oil viscosity standards.Brookfield Ametek Viscosity Standards are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify calibration of your instrument. All fluids are supplied in ½ liter (1 pint) containers complete with a certificate of calibration.Main BenefitsAccuracy: ±1% of viscosity valueExcellent temperature stabilityRecommended for use with Brookfield Ametek and most other rotational viscometersMost economicalSpecial viscosity values and temperature calibrations available upon request.
Catalog No. NC0540758
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product