Learn More
Sigma Aldrich Fine Chemicals Biosciences Viscozyme L, MFCD03101941, 50mL
Catalog No. NC1697622
Encompass
Multi-enzyme complex containing a wide range of carbohydrases, including arabanase, cellulase, β-glucanase, hemicellulase, and xylanase.
Catalog No. NC1697622 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. V201050ML
Multi-enzyme complex containing a wide range of carbohydrases, including arabanase, cellulase, β-glucanase, hemicellulase, and xylanase.