Learn More
Sigma Aldrich Fine Chemicals Biosciences ViscozymeR L cellulolytic250ML
Catalog No. 502904724
Encompass
Multi-enzyme complex containing a wide range of carbohydrases including arabanase cellulase beta-glucanase hemicellulase and xylanase
Catalog No. 50-290-4724 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. V2010250ML
Multi-enzyme complex containing a wide range of carbohydrases including arabanase cellulase beta-glucanase hemicellulase and xylanase