Learn More
Medchemexpress LLC Viscumneoside III | 118985-27-6 | 1 MG
SDP
Catalog No. 5000417390
Encompass_Preferred
Viscumneoside III is a dihydroflavone O-glycoside and a potent tyrosinase inhibitor with an IC50 of 0.5 mM. It also possesses anti-angina pectoris properties and is used as a material for cosmetics with whitening function. It shows 77% inhibition activity on tyrosinase at 1.0 mg/mL.
  • Potent tyrosinase inhibitor
  • Possesses anti-angina pectoris properties
  • Used as a material for cosmetics with whitening function
  • Shows 77% inhibition activity on tyrosinase at 1.0 mg/mL
Catalog No. 50-004-17390 Supplier Medchemexpress LLC Supplier No. HYN82231MG
Viscumneoside III is a dihydroflavone O-glycoside and a potent tyrosinase inhibitor with an IC50 of 0.5 mM. It also possesses anti-angina pectoris properties and is used as a material for cosmetics with whitening function. It shows 77% inhibition activity on tyrosinase at 1.0 mg/mL.
  • Potent tyrosinase inhibitor
  • Possesses anti-angina pectoris properties
  • Used as a material for cosmetics with whitening function
  • Shows 77% inhibition activity on tyrosinase at 1.0 mg/mL