Learn More
Abcam Visfatin Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502656177
Encompass_Preferred
Visfatin Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Visfatin.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6177 Supplier Abcam Supplier No. AB27035610X96TESTS
Visfatin Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Visfatin.
The product is subject to the following: Abcam Restricted Use Statement