Learn More
MilliporeSigma™ Supelco™ Visi-1 Single SPE Tube Processor
Catalog No. 111003771 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
Precise Flow Control
Catalog No. For Use With (Application)
11-100-3771 Precise Flow Control
1 options
Catalog No. 11-100-3771 Supplier MilliporeSigma™ Supelco™ Supplier No. 57080U
Add to Cart
Add to Cart
Provides precise flow control through a single 1, 3, or 6 mL SPE tube
  • There is no faster, more convenient, or more reliable method for processing one or a few samples
  • Simply fill the SPE tube with the appropriate solution, attach it to the Visi-1 processor, and rotate the knurled knob clockwise
  • Solution will pass through the tube in a slow, uniform rate, consistent with the most reproducible results for SPE
  • Use the plunger to expel the last drop of solvent from the tip of the tube
  • Remove the tube from the processor, introduce the next solution, and repeat the process
TRUSTED_SUSTAINABILITY
Specifications
Type SPE Tube Processor
For Use With (Application) Precise Flow Control
Quantity 1 Ea.