Learn More
Azzota Corp Visible Spectrophotometer, 2Nm Bandwidth, 320-1100Nm, 128X64 Lcd. Automatically Set Wl. Usb Connection. Can Display And Save 200 Standard Curves And 200 Groups Of Data. Pc Software Included.
Catalog No. 501613013
Encompass_Preferred
Visible Spectrophotometer, 2nm bandwidth, 320-1100nm, 128x64 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. PC scanning software included.
Catalog No. 50-161-3013 Supplier Azzota Corp Supplier No. SV1800PC
Visible Spectrophotometer, 2nm bandwidth, 320-1100nm, 128x64 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. PC scanning software included.