Learn More
MilliporeSigma™ Supelco™ Visidry™ Drying Attachment
Catalog No. 111007592 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Visiprep 12-Port Model
Visiprep 24-Port Model
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-100-7592 Visiprep 12-Port Model
11-100-7063 Visiprep 24-Port Model
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-7592 Supplier MilliporeSigma™ Supelco™ Supplier No. 57100U
Add to Cart
Add to Cart
The Visidry Drying Attachment was designed for the Visiprep Vacuum Manifold (Cat. No. 57100-U also fits the Preppy manifold).
The Visidry unit installs in minutes, dries up 12 or up to 24 SPE tubes at one time, and can be used with any inert gas supply. It is also useful for evaporating and concentrating recovered samples. Gas flow to each port can be independently adjusted.Note: The Visidry drying attachment cannot be used to dry 12, 20, or 60mL SPE tubes.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Food and Beverages
For Use With (Equipment) Visiprep 12-Port Model
Product Line Visidry
Type SPE Accessory
Visidry is a trademark of Sigma-Aldrich Co. LLC