Learn More
Sigma Aldrich Fine Chemicals Biosciences VisikolR HISTO MTM Starter
Catalog No. 502904837
Encompass
With the Visikol HISTO tissue clearing approach you are now able to rapidly and easily image tissues in 3D without damaging the underlying cellular morphology. The Visikol HISTO approach has been designed to pair with immunolabeling and is reversible so that 3D histology can be followed up with traditional 2D histology. The Visikol HISTO tissue clearing approach for 3D tissue cultures is comprised of one reagent called Visikol HISTO-M.The Visikol HISTO-M Starter Kit combines 30 mL of Visikol HISTO-M with the four buffer SOLUTIONons required to process 3D tissue cultures for immunolabeling. AAll you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of 3D cell culture models
Catalog No. 50-290-4837 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HMSK1
With the Visikol HISTO tissue clearing approach you are now able to rapidly and easily image tissues in 3D without damaging the underlying cellular morphology. The Visikol HISTO approach has been designed to pair with immunolabeling and is reversible so that 3D histology can be followed up with traditional 2D histology. The Visikol HISTO tissue clearing approach for 3D tissue cultures is comprised of one reagent called Visikol HISTO-M.The Visikol HISTO-M Starter Kit combines 30 mL of Visikol HISTO-M with the four buffer SOLUTIONons required to process 3D tissue cultures for immunolabeling. AAll you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of 3D cell culture models