Learn More
MSC Radians Lens Cleaning Liquid, 16 oz, Twist Cap, 1 ea
Catalog No. 502815354
Encompass
VisionAid lens cleaning liquids are formulated to meet all lens cleaning needs. They are safe for cleaning glass, plastic, and polycarbonate lenses, cleaning and coating them with an anti-fog, anti-static solution.
  • Cleans and coats lens with anti-fog, anti-static solution
  • Contains no silicone
  • Eliminates static with anti-fog solution
  • Complies with OSHA standards
  • Safe for cleaning glass, plastic, and polycarbonate lenses
Catalog No. 50-281-5354 Supplier MSC Supplier No. 67816629
VisionAid lens cleaning liquids are formulated to meet all lens cleaning needs. They are safe for cleaning glass, plastic, and polycarbonate lenses, cleaning and coating them with an anti-fog, anti-static solution.
  • Cleans and coats lens with anti-fog, anti-static solution
  • Contains no silicone
  • Eliminates static with anti-fog solution
  • Complies with OSHA standards
  • Safe for cleaning glass, plastic, and polycarbonate lenses