Learn More
Radians VisionAid™ SafetyClean Lens-Cleaning Towelettes
SDP
Catalog No. 19041637
Change view
Click to view available options
Product Type:
Cleaning Wipe
Catalog No. Product Type
19-041-637 Cleaning Wipe
1 options
Catalog No. 19-041-637 Supplier Radians Supplier No. LCD100
Add to Cart
Edge
Add to Cart
Antifog, antistatic premoistened towelettes
Prevent fogging and reduce static with these individually packaged, premoistened towelettes.
  • Each foil-wrapped packet contains one 5 × 8 in. (12.7 × 20.3cm) Lensclean™ tissue premoistened with Lensclean antifog, antistatic cleaning solution
  • Towelettes are silicone-free and safe for use on both glass and plastic lenses
  • Sold in dispenser packs of 100 towelettes
Specifications
Description Lens Cleaning Towelettes
Product Type Cleaning Wipe
For Use With (Equipment) Glass and plastic lenses
For Use With (Application) Prevent fogging and reduce static
No. per Pack 100 towelettes