Learn More
MSC Wells Lamont Grain Leather Driver, Goatskin, HPPE Liner, A5 Cut Protection, 12 Pairs
Catalog No. 5000687238
Encompass
These grain leather driver gloves are engineered for superior hand protection and comfort, featuring a cut-resistant liner to enhance safety in demanding work environments. The goatskin leather construction provides high tensile strength and natural abrasion resistance, while the flexible material ensures greater control when handling and gripping objects.
  • ANSI/ISEA 105-2024 cut level A5 protection
  • Abrasion level 4 and puncture level 3 rating
  • Constructed with goatskin and a fiber-glass/HPPE liner
  • Gunn cut pattern with a straight thumb
  • Double-shirred elastic wrist for a secure fit and debris protection
  • Color-coded overedge for easy size identification
  • Latex free construction
Catalog No. 50-006-87238 Supplier MSC Supplier No. 60030590
Add to Cart
Add to Cart
These grain leather driver gloves are engineered for superior hand protection and comfort, featuring a cut-resistant liner to enhance safety in demanding work environments. The goatskin leather construction provides high tensile strength and natural abrasion resistance, while the flexible material ensures greater control when handling and gripping objects.
  • ANSI/ISEA 105-2024 cut level A5 protection
  • Abrasion level 4 and puncture level 3 rating
  • Constructed with goatskin and a fiber-glass/HPPE liner
  • Gunn cut pattern with a straight thumb
  • Double-shirred elastic wrist for a secure fit and debris protection
  • Color-coded overedge for easy size identification
  • Latex free construction