Learn More
Electron Microscopy Sciences Visio-Tek™ Camera Adapters and Lens
SDP
Catalog No. 501930535
Encompass_Preferred
Kit of C-Mount Lens 0.4X/23.2 mm plus 30 and 30.5 adapters. Can be used in phototubes, trinocular tubes, or eye tubes with 23.2 mm. Connect optical device with Eye Camera.
Catalog No. 50-193-0535 Supplier Electron Microscopy Sciences Supplier No. 92113
Kit of C-Mount Lens 0.4X/23.2 mm plus 30 and 30.5 adapters. Can be used in phototubes, trinocular tubes, or eye tubes with 23.2 mm. Connect optical device with Eye Camera.