Learn More
Electron Microscopy Sciences Visio-Tek™ Eye Digital Wi-Fi Camera
SDP
Catalog No. 501930533
Encompass_Preferred
Visio-Tek Digital Microscope is a complete solution with a digital C-Mount Wi-Fi camera (VISIO-EYE) that allows you to wirelessly digitalize any traditional optical device in seconds. Compatible with all optical microscopes, the unit digitalizes images and takes measurements and photos from your standard optical device using your tablet, smartphone or PC with dedicated App and Software. The images can be displayed on different devices simultaneously. Very interesting accessory for optical microscopes. Camera only.
Catalog No. 50-193-0533 Supplier Electron Microscopy Sciences Supplier No. 92111
Visio-Tek Digital Microscope is a complete solution with a digital C-Mount Wi-Fi camera (VISIO-EYE) that allows you to wirelessly digitalize any traditional optical device in seconds. Compatible with all optical microscopes, the unit digitalizes images and takes measurements and photos from your standard optical device using your tablet, smartphone or PC with dedicated App and Software. The images can be displayed on different devices simultaneously. Very interesting accessory for optical microscopes. Camera only.