Learn More
Electron Microscopy Sciences Visio-Tek™ Scope Digital Wi-Fi Camera with Lens and Adapters
SDP
Catalog No. 501930531
Encompass_Preferred
Visio-Tek Digital Microscope is a complete solution with a digital C-Mount Wi-Fi camera (VISIO-EYE) that allows you to wirelessly digitalize any traditional optical device in seconds. Compatible with all optical microscopes, the unit digitalizes images and takes measurements and photos from your standard optical device using your tablet, smartphone or PC with dedicated App and Software. The images can be displayed on different devices simultaneously. Very interesting accessory for optical microscopes. Kit includes Cat. #92111 Wi-Fi Camera, Cat. #92113 Lens & Adapters and Cat. #92114 Spacer Rings.
Catalog No. 50-193-0531 Supplier Electron Microscopy Sciences Supplier No. 92109
Visio-Tek Digital Microscope is a complete solution with a digital C-Mount Wi-Fi camera (VISIO-EYE) that allows you to wirelessly digitalize any traditional optical device in seconds. Compatible with all optical microscopes, the unit digitalizes images and takes measurements and photos from your standard optical device using your tablet, smartphone or PC with dedicated App and Software. The images can be displayed on different devices simultaneously. Very interesting accessory for optical microscopes. Kit includes Cat. #92111 Wi-Fi Camera, Cat. #92113 Lens & Adapters and Cat. #92114 Spacer Rings.