Learn More
MilliporeSigma™ Supelco™ Visiprep™ 5-port Flask DL (Disposable Liner) Vacuum Manifold
Catalog No. 111006048 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
Food and Beverages
Catalog No. For Use With (Application)
11-100-6048 Food and Beverages
1 options
Catalog No. 11-100-6048 Supplier MilliporeSigma™ Supelco™ Supplier No. 57101U
Add to Cart
Add to Cart
The Visiprep 5-Port Flask Vacuum Manifold enables analysts using Supelco solid phase extraction tubes to simultaneously prepare up to 5 samples.
Unlike conventional vacuum manifolds, the Visiprep 5-Port Flask Manifold allows users to collect their SPE eluate directly into 50 mL round or flat bottom flasks for direct Rotovap evaporation. The manifold consists of a chemical resistant 5-port cover (DL or Standard available), gasket, base, a glass basin, vacuum gauge and bleed valve, 5 flow control valves, 5 replaceable solvent guide needles, and a base plate that supports up to five 50 mL round or flat bottom flasks. Each port on both the standard and DL Visiprep models are equipped with flow control valves. Recommended Flasks: Aldrich™ single-neck flask, 50 mL, joint: ST/NS 24/40. Round Bottom (Cat. No. Z414484). Flat Bottom (Cat. No. Z418773).
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Food and Beverages
Product Line Visiprep
Type SPE Accessory
Visiprep is a trademark of Sigma-Aldrich Co. LLC