Learn More
MilliporeSigma™ Supelco™ Visiprep™ Manifold Collection Rack
Catalog No. 111003912 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Visiprep 12-Port Model
Visiprep 24-Port Model
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-100-3912 Visiprep 12-Port Model
11-100-3829 Visiprep 24-Port Model
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-3912 Supplier MilliporeSigma™ Supelco™ Supplier No. 57037
Add to Cart
Add to Cart
For use with Visiprep Vacuum Manifold
TRUSTED_SUSTAINABILITY
Specifications
Description Included with standard and DL Visiprep Vacuum manifold models
For Use With (Application) Food and Beverages
For Use With (Equipment) Visiprep 12-Port Model
Product Line Visiprep
Type SPE Accessory
Includes Base, 3 support rods, center plate, plate for 10 mm test tubes, 12 retaining clips
Visiprep is a trademark of Sigma-Aldrich Co. LLC