Learn More
Crescent Chemical Co Inc VISKING DIALYSIS TUBING
SDP
Supplier:  Crescent Chemical Co Inc SE44110.04
Encompass_Preferred
VISKING dialysis tubing diameter 16 mm Packed dry , made from regenerated cellulose (RC). With glycerol as protection for embrittlement, which can be easily removed by soaking in water. Contain low level of heavy metal and sulfide impurities. Highly resistant against che
Catalog No. 50-248-842