Learn More
Crescent Chemical Co Inc VISKING DIALYSIS TUBING REG
SDP
Supplier:  Crescent Chemical Co Inc SE44114.01
Encompass_Preferred
VISKING dialysis tubing regenerated cellulose, diameter 21 mm Packed dry , made from regenerated cellulose (RC). With glycerol as protection for embrittlement, which can be easily removed by soaking in water. Contain low level of heavy metal and sulfide impurities. Highly
Catalog No. 50-248-843