Learn More
Medchemexpress LLC HY-10440 50mg , Vismodegib CAS:879085-55-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1044050MG
Encompass_Preferred
HY-10440 50mg Vismodegib CAS: 879085-55-9 Vismodegib (GDC-0449) is an orally active hedgehog pathway inhibitor with an IC50 of 3 nM. Vismodegib also inhibits P-gp, ABCG2 with IC50 values of 3.0 μM and 1.4 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9915