Learn More
Selleck Chemical LLC Vismodegib (GDC-0449) 50mg 879085-55-9
SDP
Supplier:  Selleck Chemical LLC S108250MG
Encompass_Preferred
Vismodegib (GDC-0449) is a potent, novel and specific hedgehog inhibitor with IC50 of 3 nM and also inhibits P-gp with IC50 of 3.0 ?M in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-779-8